accessibility menu, dialog, popup

UserName

CH3(S)(S)HOH3CCH3

CAS RN 933-48-2

IUPAC Name: (1S,5S)-3,3,5-trimethylcyclohexan-1-ol
Synonyms: (1S,5S)-3,3,5-trimethylcyclohexan-1-ol
Molecular Weight (g/mol): 142.24
Molecular Formula: C9H18O
InChi Key: BRRVXFOKWJKTGG-SFYZADRCSA-N
SMILES: C[C@@H]1C[C@H](O)CC(C)(C)C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results