missing translation for 'onlineSavingsMsg'
Learn More

cis-3,3,5-Trimethylcyclohexanol (contains ca. 20% trans- isomer) 80.0+%, TCI America™

Catalog No. T269325G
Change view
Click to view available options
Quantity:
25 g
400 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T269325G 25 g
T2693400G 400 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T269325G Supplier TCI America Supplier No. T269325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 116-02-9
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
MDL Number MFCD00019378
InChI Key BRRVXFOKWJKTGG-UHFFFAOYSA-N
Synonym 3,3,5-trimethylcyclohexanol, cyclonol, homomenthol, cyclohexanol, 3,3,5-trimethyl, isophorol, dihydro, trimethylcyclohexanol, dihydroisophorol, 3,3,5-trimethyl-1-cyclohexanol, 3,5,5-trimethylcyclohexanol, 1-methyl-3-dimethylcyclohexanol-5
PubChem CID 8298
ChEBI CHEBI:59065
IUPAC Name 3,3,5-trimethylcyclohexan-1-ol
SMILES CC1CC(CC(C1)(C)C)O

Specifications

Melting Point 34°C
Color White
Boiling Point 86°C
Quantity 25 g
Formula Weight 142.24
Percent Purity ≥80.0% (GC)
Physical Form Crystals
Chemical Name or Material cis-3,3,5-Trimethylcyclohexanol (contains ca. 20% trans- isomer)
EINECSNumber (3)-2294
RTECSNumber GW0875000
TSCA Yes