accessibility menu, dialog, popup

UserName

CAS RN 116-02-9

IUPAC Name: 3,3,5-trimethylcyclohexan-1-ol
Synonyms: 3,3,5-trimethylcyclohexanol trimethylcyclohexanol dihydroisophorol homomenthol
Molecular Weight (g/mol): 142.24
Molecular Formula: C9H18O
InChi Key: BRRVXFOKWJKTGG-UHFFFAOYNA-N
SMILES: CC1CC(O)CC(C)(C)C1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)