missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Octene 95.0+%, TCI America™

Catalog No. O00985ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O00985ML 5 mL
O00981ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O00985ML Supplier TCI America Supplier No. O00985ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7642-04-8
Molecular Formula C8H16
Molecular Weight (g/mol) 112.22
MDL Number MFCD00065146
InChI Key ILPBINAXDRFYPL-HWKANZROSA-N
PubChem CID 5364448
IUPAC Name (E)-oct-2-ene
SMILES CCCCCC=CC

Specifications

Color Colorless
Boiling Point 126°C
Quantity 5 mL
UN Number 3295
Formula Weight 112.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-2-Octene
EINECSNumber (2)-0024
TSCA No