accessibility menu, dialog, popup

UserName

H3CCH3(E)

CAS RN 7642-04-8

IUPAC Name: (2E)-oct-2-ene
Synonyms: 2-octene (cis) 2-octene (trans)
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: ILPBINAXDRFYPL-HWKANZROSA-N
SMILES: CCCCC\C=C\C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results