missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Heptene 95.0+%, TCI America™

Catalog No. p-7130530
Change view
Click to view available options
Quantity:
1 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04111ML 1 mL
H041110ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H04111ML Supplier TCI America Supplier No. H04111ML

Chemical Identifiers

CAS 6443-92-1
Molecular Formula C7H14
Molecular Weight (g/mol) 98.189
MDL Number MFCD00063976
InChI Key OTTZHAVKAVGASB-HYXAFXHYSA-N
PubChem CID 643836
IUPAC Name (Z)-hept-2-ene
SMILES CCCCC=CC

Specifications

Color Colorless
Boiling Point 99°C
Quantity 1 mL
UN Number 3295
Formula Weight 98.19
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-2-Heptene