CAS RN 6443-92-1
IUPAC Name:
(2Z)-hept-2-ene
Synonyms:
2-heptene (cis)
Molecular Weight (g/mol):
98.19
Molecular Formula:
C7H14
InChi Key:
OTTZHAVKAVGASB-HYXAFXHYSA-N
SMILES:
CCCC\C=C/C
1
–
1
of
1
results
1
cis-2-Heptene 95.0+%, TCI America™
CAS: 6443-92-1 Molecular Formula: C7H14 Molecular Weight (g/mol): 98.189 MDL Number: MFCD00063976 InChI Key: OTTZHAVKAVGASB-HYXAFXHYSA-N PubChem CID: 643836 IUPAC Name: (Z)-hept-2-ene SMILES: CCCCC=CC