missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-2-(Boc-amino)cyclopropanemethanol, 97%
CAS: 170299-61-3 | C9H17NO3 | 187.239 g/mol
$457.88 - $1,290.05
Chemical Identifiers
| CAS | 170299-61-3 |
|---|---|
| Molecular Formula | C9H17NO3 |
| Molecular Weight (g/mol) | 187.239 |
| MDL Number | MFCD03844611 |
| InChI Key | OCKKMJSVLVALMF-RNFRBKRXSA-N |
| Synonym | tert-butyl cis-2-hydroxymethyl cyclopropylcarbamate, cis-1-boc-amino-2-hydroxymethyl cyclopropane, tert-butyl cis-2-hydroxymethyl-cyclopropylcarbamate, tert-butyl 1r,2s-2-hydroxymethyl cyclopropyl carbamate, 2-methyl-2-propanyl 1r,2s-2-hydroxymethyl cyclopropyl carbamate, tert-butyl n-1r,2s-2-hydroxymethyl cyclopropyl carbamate, carbamic acid, 2-hydroxymethyl cyclopropyl-, 1,1-dimethylethyl ester, 1r, carbamic acid, 2-hydroxymethyl cyclopropyl-, 1,1-dimethylethyl ester, cis |
| PubChem CID | 24720933 |
| IUPAC Name | tert-butyl N-[(1R,2S)-2-(hydroxymethyl)cyclopropyl]carbamate |
| SMILES | CC(C)(C)OC(=O)NC1CC1CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAH62795MD
|
N/A | 250 mg |
Each for $457.88
|
|
|||||
|
AAH6279503
|
N/A | 1 g |
Each for $1,290.05
|
|
|||||
Description
cis-2-(Boc-amino)cyclopropanemethanol is employed as a intermediate for chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationscis-2-(Boc-amino)cyclopropanemethanol is employed as a intermediate for chemical research.
Notes
Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
| 170299-61-3 | |
| 187.239 | |
| OCKKMJSVLVALMF-RNFRBKRXSA-N | |
| 24720933 | |
| CC(C)(C)OC(=O)NC1CC1CO |
| C9H17NO3 | |
| MFCD03844611 | |
| tert-butyl cis-2-hydroxymethyl cyclopropylcarbamate, cis-1-boc-amino-2-hydroxymethyl cyclopropane, tert-butyl cis-2-hydroxymethyl-cyclopropylcarbamate, tert-butyl 1r,2s-2-hydroxymethyl cyclopropyl carbamate, 2-methyl-2-propanyl 1r,2s-2-hydroxymethyl cyclopropyl carbamate, tert-butyl n-1r,2s-2-hydroxymethyl cyclopropyl carbamate, carbamic acid, 2-hydroxymethyl cyclopropyl-, 1,1-dimethylethyl ester, 1r, carbamic acid, 2-hydroxymethyl cyclopropyl-, 1,1-dimethylethyl ester, cis | |
| tert-butyl N-[(1R,2S)-2-(hydroxymethyl)cyclopropyl]carbamate |
Specifications
| 170299-61-3 | |
| MFCD03844611 | |
| OCKKMJSVLVALMF-RNFRBKRXSA-N | |
| tert-butyl N-[(1R,2S)-2-(hydroxymethyl)cyclopropyl]carbamate | |
| 24720933 | |
| 97% |
| C9H17NO3 | |
| tert-butyl cis-2-hydroxymethyl cyclopropylcarbamate, cis-1-boc-amino-2-hydroxymethyl cyclopropane, tert-butyl cis-2-hydroxymethyl-cyclopropylcarbamate, tert-butyl 1r,2s-2-hydroxymethyl cyclopropyl carbamate, 2-methyl-2-propanyl 1r,2s-2-hydroxymethyl cyclopropyl carbamate, tert-butyl n-1r,2s-2-hydroxymethyl cyclopropyl carbamate, carbamic acid, 2-hydroxymethyl cyclopropyl-, 1,1-dimethylethyl ester, 1r, carbamic acid, 2-hydroxymethyl cyclopropyl-, 1,1-dimethylethyl ester, cis | |
| CC(C)(C)OC(=O)NC1CC1CO | |
| 187.239 | |
| 187.23 | |
| cis-2-(Boc-amino)cyclopropanemethanol |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only