missing translation for 'onlineSavingsMsg'
Learn More

cis-2-(Boc-amino)cyclopropanemethanol, 97%

CAS: 170299-61-3 | C9H17NO3 | 187.239 g/mol

Supplier:  Thermo Scientific Chemicals H6279503

Quantity 1 g

 View more versions of this product

Catalog No. AAH6279503


Only null left

Description

Description

cis-2-(Boc-amino)cyclopropanemethanol is employed as a intermediate for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-2-(Boc-amino)cyclopropanemethanol is employed as a intermediate for chemical research.

Notes
Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

170299-61-3
187.239
OCKKMJSVLVALMF-RNFRBKRXSA-N
24720933
CC(C)(C)OC(=O)NC1CC1CO
C9H17NO3
MFCD03844611
tert-butyl cis-2-hydroxymethyl cyclopropylcarbamate, cis-1-boc-amino-2-hydroxymethyl cyclopropane, tert-butyl cis-2-hydroxymethyl-cyclopropylcarbamate, tert-butyl 1r,2s-2-hydroxymethyl cyclopropyl carbamate, 2-methyl-2-propanyl 1r,2s-2-hydroxymethyl cyclopropyl carbamate, tert-butyl n-1r,2s-2-hydroxymethyl cyclopropyl carbamate, carbamic acid, 2-hydroxymethyl cyclopropyl-, 1,1-dimethylethyl ester, 1r, carbamic acid, 2-hydroxymethyl cyclopropyl-, 1,1-dimethylethyl ester, cis
tert-butyl N-[(1R,2S)-2-(hydroxymethyl)cyclopropyl]carbamate

Specifications

cis-2-(Boc-amino)cyclopropanemethanol
C9H17NO3
tert-butyl cis-2-hydroxymethyl cyclopropylcarbamate, cis-1-boc-amino-2-hydroxymethyl cyclopropane, tert-butyl cis-2-hydroxymethyl-cyclopropylcarbamate, tert-butyl 1r,2s-2-hydroxymethyl cyclopropyl carbamate, 2-methyl-2-propanyl 1r,2s-2-hydroxymethyl cyclopropyl carbamate, tert-butyl n-1r,2s-2-hydroxymethyl cyclopropyl carbamate, carbamic acid, 2-hydroxymethyl cyclopropyl-, 1,1-dimethylethyl ester, 1r, carbamic acid, 2-hydroxymethyl cyclopropyl-, 1,1-dimethylethyl ester, cis
CC(C)(C)OC(=O)NC1CC1CO
1 g
24720933
97%
170299-61-3
MFCD03844611
OCKKMJSVLVALMF-RNFRBKRXSA-N
tert-butyl N-[(1R,2S)-2-(hydroxymethyl)cyclopropyl]carbamate
187.239
187.23
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions