missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-2-Amino-1-cyclopentanecarboxylic acid, 98+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 218.0°C to 220.0°C |
| Assay Percent Range | 98+% |
| Packaging | Glass Bottle |
| Molecular Formula | C6H11NO2 |
| Solubility Information | Solubility in water: soluble in water. Other solubilities: soluble in methanol |
| Quantity | 2 g |
| Molecular Weight (g/mol) | 129.16 |
| Formula Weight | 129.16 |
| Percent Purity | ≥98% |
| Chemical Name or Material | cis-2-Amino-1-cyclopentanecarboxylic acid, 98+% |
RUO – Research Use Only