accessibility menu, dialog, popup

UserName

CAS RN 37910-65-9

NH2OHO

CAS RN 37910-65-9

IUPAC Name: 2-aminocyclopentane-1-carboxylic acid
Synonyms: 2-aminocyclopentane-1-carboxylic acid
Molecular Weight (g/mol): 129.16
Molecular Formula: C6H11NO2
InChi Key: JWYOAMOZLZXDER-UHFFFAOYNA-N
SMILES: NC1CCCC1C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)