missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Amino-1-cyclopentanecarboxylic acid, 98+%

CAS: 37910-65-9 | C6H11NO2 | 129.159 g/mol

$737.24 - $737.24

Chemical Identifiers

CAS 37910-65-9
MDL Number MFCD00143982
InChI Key JWYOAMOZLZXDER-UHNVWZDZSA-N
Synonym 1r,2s-2-aminocyclopentanecarboxylic acid, cispentacin, cis-2-amino-1-cyclopentanecarboxylic acid, --cispentacin, cis-pentacin, antibiotic fr 109615, 1r,2s-2-aminocyclopentane-1-carboxylic acid, --1r,2s-cispentacin, +/--cispentacin, cispentacin,-
PubChem CID 73305
IUPAC Name (1R,2S)-2-aminocyclopentane-1-carboxylic acid
SMILES C1CC(C(C1)N)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC277740020
SDS View Documents
Thermo Scientific Chemicals
277740020
2 g
Each for $737.24
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

37910-65-9
JWYOAMOZLZXDER-UHNVWZDZSA-N
73305
C1CC(C(C1)N)C(=O)O
MFCD00143982
1r,2s-2-aminocyclopentanecarboxylic acid, cispentacin, cis-2-amino-1-cyclopentanecarboxylic acid, --cispentacin, cis-pentacin, antibiotic fr 109615, 1r,2s-2-aminocyclopentane-1-carboxylic acid, --1r,2s-cispentacin, +/--cispentacin, cispentacin,-
(1R,2S)-2-aminocyclopentane-1-carboxylic acid

Specifications

218.0°C to 220.0°C
98+%
Glass Bottle
1r,2s-2-aminocyclopentanecarboxylic acid, cispentacin, cis-2-amino-1-cyclopentanecarboxylic acid, --cispentacin, cis-pentacin, antibiotic fr 109615, 1r,2s-2-aminocyclopentane-1-carboxylic acid, --1r,2s-cispentacin, +/--cispentacin, cispentacin,-
JWYOAMOZLZXDER-UHNVWZDZSA-N
(1R,2S)-2-aminocyclopentane-1-carboxylic acid
129.16
129.16
cis-2-Amino-1-cyclopentanecarboxylic acid, 98+%
37910-65-9
C6H11NO2
MFCD00143982
Solubility in water: soluble in water. Other solubilities: soluble in methanol
C1CC(C(C1)N)C(=O)O
2 g
73305
≥98%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only