missing translation for 'onlineSavingsMsg'
Learn More

cis-2,6-Dimethylpiperidine 99.0+%, TCI America™

Catalog No. p-7126632
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D119825ML 25 mL
D1198100ML 100 mL
D1198500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D119825ML Supplier TCI America Supplier No. D119825ML

Chemical Identifiers

CAS 766-17-6
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.204
MDL Number MFCD00066304
InChI Key SDGKUVSVPIIUCF-KNVOCYPGSA-N
Synonym cis-2,6-dimethylpiperidine, 2r,6s-2,6-dimethylpiperidine, cis-hexahydro-2,6-lutidine, cis-2,6-lupetidine, piperidin, 2e,6e-dimethyl, cis-2,6-dimethyl-piperidine, cis-lupetidine, cis-nanophine, prestwick0_000666, prestwick1_000666
PubChem CID 641768
IUPAC Name (2R,6S)-2,6-dimethylpiperidine
SMILES CC1CCCC(N1)C

Specifications

Color Yellow
Boiling Point 127°C
Quantity 25 mL
UN Number 2733
Formula Weight 113.20
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material cis-2,6-Dimethylpiperidine
EINECSNumber (5)-3728
RTECSNumber OK5775000
TSCA No