accessibility menu, dialog, popup

UserName

CH3H3CHN

CAS RN 766-17-6

IUPAC Name: 2,6-dimethylpiperidine
Synonyms: 2,6-dimethylpiperidine 2,6-dimethylpiperidine (cis-)
Molecular Weight (g/mol): 113.2
Molecular Formula: C7H15N
InChi Key: SDGKUVSVPIIUCF-UHFFFAOYNA-N
SMILES: CC1CCCC(C)N1
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results