Learn More
cis-2,3-Epoxybutane, 98%, Thermo Scientific Chemicals
$177.36 - $177.36
Chemical Identifiers
CAS | 1758-33-4 |
---|---|
Molecular Formula | C4H8O |
Molecular Weight (g/mol) | 72.11 |
MDL Number | MFCD00005128 |
InChI Key | PQXKWPLDPFFDJP-ZXZARUISSA-N |
Synonym | cis-2,3-epoxybutane, cis-2-butene oxide, cis-2,3-dimethyloxirane, oxirane, 2,3-dimethyl-, cis, cis-2-butene epoxide, cis-2-butylene oxide, butane, 2,3-epoxy-, cis, ccris 3758, 2r,3s-2,3-dimethyloxirane, meso-2,3-epoxybutane |
PubChem CID | 92162 |
IUPAC Name | (2R,3S)-2,3-dimethyloxirane |
SMILES | C[C@H]1O[C@H]1C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1032006
|
Thermo Scientific Chemicals
L1032006 |
5 g |
Each for $177.36
|
|
|||||
Description
Used in the enantioselective methanolysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed in the enantioselective methanolysis.
Solubility
Soluble in water, 2.848e+004 mg/L.
Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture. Store at 4°C.
Chemical Identifiers
1758-33-4 | |
72.11 | |
PQXKWPLDPFFDJP-ZXZARUISSA-N | |
92162 | |
C[C@H]1O[C@H]1C |
C4H8O | |
MFCD00005128 | |
cis-2,3-epoxybutane, cis-2-butene oxide, cis-2,3-dimethyloxirane, oxirane, 2,3-dimethyl-, cis, cis-2-butene epoxide, cis-2-butylene oxide, butane, 2,3-epoxy-, cis, ccris 3758, 2r,3s-2,3-dimethyloxirane, meso-2,3-epoxybutane | |
(2R,3S)-2,3-dimethyloxirane |
Specifications
1758-33-4 | |
0.826 | |
−21°C (−5°F) | |
1.383 | |
5 g | |
79770 | |
Soluble in water,2.848e+004mg/L. | |
C[C@H]1O[C@H]1C | |
72.11 | |
72.11 | |
cis-2,3-Epoxybutane |
-84°C | |
60°C to 61°C | |
C4H8O | |
MFCD00005128 | |
UN3271 | |
cis-2,3-epoxybutane, cis-2-butene oxide, cis-2,3-dimethyloxirane, oxirane, 2,3-dimethyl-, cis, cis-2-butene epoxide, cis-2-butylene oxide, butane, 2,3-epoxy-, cis, ccris 3758, 2r,3s-2,3-dimethyloxirane, meso-2,3-epoxybutane | |
PQXKWPLDPFFDJP-ZXZARUISSA-N | |
(2R,3S)-2,3-dimethyloxirane | |
92162 | |
98% |
Safety and Handling
GHS H Statement
H225-H341-H315-H319-H335
Highly flammable liquid and vapor.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H225-H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber : 000-000-0
RTECSNumber : EK3855020
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only