accessibility menu, dialog, popup

UserName

CH3(R)O(S)H3C

CAS RN 1758-33-4

IUPAC Name: (2R,3S)-2,3-dimethyloxirane
Synonyms: (2R,3S)-2,3-dimethyloxirane
Molecular Weight (g/mol): 72.11
Molecular Formula: C4H8O
InChi Key: PQXKWPLDPFFDJP-ZXZARUISSA-N
SMILES: C[C@H]1O[C@H]1C
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)