missing translation for 'onlineSavingsMsg'
Learn More

cis-11-Eicosenoic acid, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014604
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014604 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014604 Supplier Merck Emd Millipore Supplier No. 44878100MG
Only null left

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 5561-99-9
Density 0.883 g/mL (at 25°C)
Flash Point >113°C
Physical Form Neat
Linear Formula CH3(CH2)7 CH=CH(CH2)9 CO2 H
MDL Number MFCD00010506
Melting Point 23°C to 24°C
Synonym Gondoic acid
Type Analytical Standard
Quantity 100 mg
Formula Weight 310.51
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less