missing translation for 'onlineSavingsMsg'
Learn More

cis-1,4-Dichloro-2-butene, 95%

Catalog No. AAA1895614
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1895614 25 g
AAA1895622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1895614 Supplier Thermo Scientific Chemicals Supplier No. A1895614

CAS: 1476-11-5 | C4H6Cl2 | 124.992 g/mol

It is employed as an intermediate for the production of other chemicals such as in the production of chloroprene. It is also used as a starting material in the production of adiponitrile, butane-1,4-diol, and tetrahydrofuran. It is also used in the preparation of functionalized 3,5-disubstituted cyclopent-2-enones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an intermediate for the production of other chemicals such as in the production of chloroprene. It is also used as a starting material in the production of adiponitrile, butane-1,4-diol, and tetrahydrofuran. It is also used in the preparation of functionalized 3,5-disubstituted cyclopent-2-enones.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol, ether, acetone and benzene.

Notes
Moisture & Light Sensitive. Store away from oxidizing agents, heat, water/moisture and light. Store under inert gas. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.

Chemical Identifiers

CAS 1476-11-5
Molecular Formula C4H6Cl2
MDL Number MFCD00062950
InChI Key FQDIANVAWVHZIR-UPHRSURJSA-N
Synonym cis-1,4-dichloro-2-butene, z-1,4-dichlorobut-2-ene, 1,4-dichloro-cis-2-butene, 2-butene, 1,4-dichloro-, z, cis-1,4-dichlorobut-2-ene, unii-bqs2752i90, ccris 2651, cis 1,4-dichlorobutene, 2-butene, 1,4-dichloro-, cis, cis-2,3-dichlorobut-2-ene
PubChem CID 6432115
IUPAC Name (Z)-1,4-dichlorobut-2-ene

Specifications

Melting Point -48°C
Density 1.188
Boiling Point 151°C to 153°C
Flash Point 55°C (132°F)
Refractive Index 1.49
Quantity 25 g
UN Number UN3390
Beilstein 1719692
Sensitivity Light and moisture sensitive
Solubility Information Not miscible or difficult to mix in water. Soluble in alcohol,ether,acetone and benzene.
SMILES C(C=CCCl)Cl
Molecular Weight (g/mol) 124.992
Formula Weight 125
Percent Purity 95%
Chemical Name or Material cis-1,4-Dichloro-2-butene
Show More Show Less
Hazard Category H226-H301+H311-H314-H330-H335
Hazard Statement GHS H Statement
H300-H310-H330-H350-H314-H318-H226
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause cancer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, CORROSIVE, N.O.S.
EINECSNumber 216-021-5
RTECSNumber EM4900000
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only