accessibility menu, dialog, popup

UserName

ClCl(Z)

CAS RN 1476-11-5

IUPAC Name: (2Z)-1,4-dichlorobut-2-ene
Synonyms: 1,4-dichloro-2-butene(cis)
Molecular Weight (g/mol): 124.99
Molecular Formula: C4H6Cl2
InChi Key: FQDIANVAWVHZIR-UPHRSURJSA-N
SMILES: ClC\C=C/CCl
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)