missing translation for 'onlineSavingsMsg'
Learn More

cis-1,4-Dichloro-2-butene, 95%

Catalog No. p-4509647
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC187600250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC187600250 Supplier Thermo Scientific Chemicals Supplier No. 187600250

CAS: 1476-11-5 | C4H6Cl2 | 125 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1476-11-5
Molecular Formula C4H6Cl2
Molecular Weight (g/mol) 125
InChI Key FQDIANVAWVHZIR-UPHRSURJSA-N
Synonym cis-1,4-dichloro-2-butene, z-1,4-dichlorobut-2-ene, 1,4-dichloro-cis-2-butene, 2-butene, 1,4-dichloro-, z, cis-1,4-dichlorobut-2-ene, unii-bqs2752i90, ccris 2651, cis 1,4-dichlorobutene, 2-butene, 1,4-dichloro-, cis, cis-2,3-dichlorobut-2-ene
PubChem CID 6432115
IUPAC Name (Z)-1,4-dichlorobut-2-ene
SMILES C(C=CCCl)Cl

Specifications

Melting Point -48.0°C
Color Colorless to Yellow
Density 1.1800g/mL
Boiling Point 152.0°C (758.0 mmHg)
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4884 to 1.4914
Linear Formula ClCH2CH=CHCH2Cl
Quantity 25 mL
Beilstein ,III,743
Fieser 10,134; 01,519; 02,229; 03,167; 04,268; 05,357; 06,304; 10,221; 13,152; 17,270
Specific Gravity 1.18
Formula Weight 125
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material cis-1, 4-Dichloro-2-butene
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 216-021-5

RUO – Research Use Only