missing translation for 'onlineSavingsMsg'
Learn More

cis-1,4-Diacetoxy-2-butene 95.0+%, TCI America™

Catalog No. D135825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D135825ML 25 mL
D1358500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D135825ML Supplier TCI America Supplier No. D135825ML

Chemical Identifiers

CAS 25260-60-0
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00059339,MFCD00077968
InChI Key VZUAUHWZIKOMFC-ARJAWSKDSA-N
Synonym cis-1,4-diacetoxy-2-butene, z-but-2-ene-1,4-diyl diacetate, 2z-but-2-ene-1,4-diyl diacetate, cis-2-butene-1,4-diol diacetate, 2z-4-acetyloxybut-2-enyl acetate, 2z-4-acetyloxy but-2-en-1-yl acetate, cis-1,4-diacetoxy-2-buten, z-1,4-diacetoxy-2-butene, cis-2-buten-1,4-diol diacetate, z-but-2-ene-1,4-diyldiacetate
PubChem CID 643799
IUPAC Name (2Z)-4-(acetyloxy)but-2-en-1-yl acetate
SMILES CC(=O)OC\C=C/COC(C)=O

Specifications

Color Yellow
Boiling Point 110°C
Quantity 25 mL
Formula Weight 172.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-1,4-Diacetoxy-2-butene
EINECSNumber (2)-3305
TSCA No