accessibility menu, dialog, popup

UserName

CH3OOOH3CO(Z)

CAS RN 25260-60-0

IUPAC Name: (2Z)-4-(acetyloxy)but-2-en-1-yl acetate
Synonyms: (2Z)-4-(acetyloxy)but-2-en-1-yl acetate
Molecular Weight (g/mol): 172.18
Molecular Formula: C8H12O4
InChi Key: VZUAUHWZIKOMFC-ARJAWSKDSA-N
SMILES: CC(=O)OC\C=C/COC(C)=O
Boiling Point 
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)