missing translation for 'onlineSavingsMsg'
Learn More

cis-1,4-Diacetoxy-2-butene, 96%

Catalog No. AAL1302409
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1302409 10 g
AAL1302418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1302409 Supplier Thermo Scientific Chemicals Supplier No. L1302409
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 25260-60-0 | C8H12O4 | 172.18 g/mol

cis-1,4-Diacetoxy-2-butene is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-1,4-Diacetoxy-2-butene is used as an organic chemical synthesis intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 25260-60-0
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00059339,MFCD00077968
InChI Key VZUAUHWZIKOMFC-ARJAWSKDSA-N
Synonym cis-1,4-diacetoxy-2-butene, z-but-2-ene-1,4-diyl diacetate, 2z-but-2-ene-1,4-diyl diacetate, cis-2-butene-1,4-diol diacetate, 2z-4-acetyloxybut-2-enyl acetate, 2z-4-acetyloxy but-2-en-1-yl acetate, cis-1,4-diacetoxy-2-buten, z-1,4-diacetoxy-2-butene, cis-2-buten-1,4-diol diacetate, z-but-2-ene-1,4-diyldiacetate
PubChem CID 643799
IUPAC Name [(Z)-4-acetyloxybut-2-enyl] acetate
SMILES CC(=O)OC\C=C/COC(C)=O

Specifications

Density 1.09
Boiling Point 120°C to 121°C (18 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4416
Quantity 10 g
Beilstein 1724379
Solubility Information Not miscible in water.
Formula Weight 172.18
Percent Purity 96%
Chemical Name or Material cis-1,4-Diacetoxy-2-butene
Hazard Category H315
Precautionary Statement P264b-P280-P302+P352-P332+P313-P362
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only