missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-1,4-Cyclohexanedicarboxylic Acid 90.0+%, TCI America™
$175.22 - $479.06
Chemical Identifiers
| CAS | 619-81-8 |
|---|---|
| Molecular Formula | C8H12O4 |
| Molecular Weight (g/mol) | 172.18 |
| MDL Number | MFCD00064950 |
| InChI Key | PXGZQGDTEZPERC-UHFFFAOYSA-N |
| PubChem CID | 14106 |
| IUPAC Name | cyclohexane-1,4-dicarboxylic acid |
| SMILES | C1CC(CCC1C(=O)O)C(=O)O |
Chemical Identifiers
| 619-81-8 | |
| 172.18 | |
| PXGZQGDTEZPERC-UHFFFAOYSA-N | |
| cyclohexane-1,4-dicarboxylic acid |
| C8H12O4 | |
| MFCD00064950 | |
| 14106 | |
| C1CC(CCC1C(=O)O)C(=O)O |
Specifications
| 619-81-8 | |
| White | |
| MFCD00064950 | |
| PXGZQGDTEZPERC-UHFFFAOYSA-N | |
| cyclohexane-1,4-dicarboxylic acid | |
| 14106 | |
| ≥90.0% (GC) | |
| cis-1,4-Cyclohexanedicarboxylic Acid |
| 168°C | |
| C8H12O4 | |
| 25 g | |
| C1CC(CCC1C(=O)O)C(=O)O | |
| 172.18 | |
| 172.18 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-2423&(3)-2446
TSCA : No