missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-1,4-Cyclohexanedicarboxylic Acid 90.0+%, TCI America™
Supplier: TCI America C0789100G
Specifications
| cis-1,4-Cyclohexanedicarboxylic Acid | |
| 168°C | |
| C8H12O4 | |
| 100 g | |
| C1CC(CCC1C(=O)O)C(=O)O | |
| 172.18 | |
| 172.18 | |
| Crystalline Powder |
| 619-81-8 | |
| White | |
| MFCD00064950 | |
| PXGZQGDTEZPERC-UHFFFAOYSA-N | |
| cyclohexane-1,4-dicarboxylic acid | |
| 14106 | |
| ≥90.0% (GC) |
Chemical Identifiers
| 619-81-8 | |
| 172.18 | |
| PXGZQGDTEZPERC-UHFFFAOYSA-N | |
| cyclohexane-1,4-dicarboxylic acid |
| C8H12O4 | |
| MFCD00064950 | |
| 14106 | |
| C1CC(CCC1C(=O)O)C(=O)O |
Safety and Handling
EINECSNumber : (3)-2423&(3)-2446
TSCA : No