missing translation for 'onlineSavingsMsg'
Learn More

cis-1,3-Dibenzyl-2-oxo-4,5-imidazolidinedicarboxylic Acid 98.0+%, TCI America™

Catalog No. D330925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D330925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D330925G Supplier TCI America Supplier No. D330925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51591-75-4
Molecular Formula C19H18N2O5
Molecular Weight (g/mol) 354.36
MDL Number MFCD06798134
InChI Key QSMUFXXTSUEZJA-UHFFFAOYNA-N
PubChem CID 9841512
IUPAC Name 1,3-dibenzyl-2-oxoimidazolidine-4,5-dicarboxylic acid
SMILES OC(=O)C1C(N(CC2=CC=CC=C2)C(=O)N1CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 175°C
Color White-Yellow
Quantity 25 g
Formula Weight 354.36
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material cis-1,3-Dibenzyl-2-oxo-4,5-imidazolidinedicarboxylic Acid
EINECSNumber (5)-3591
TSCA No