accessibility menu, dialog, popup

UserName

CAS RN 51591-75-4

IUPAC Name: 1,3-dibenzyl-2-oxoimidazolidine-4,5-dicarboxylic acid
Synonyms: 1,3-dibenzyl-2-oxoimidazolidine-4,5-dicarboxylic acid
Molecular Weight (g/mol): 354.36
Molecular Formula: C19H18N2O5
InChi Key: QSMUFXXTSUEZJA-UHFFFAOYNA-N
SMILES: OC(=O)C1C(N(CC2=CC=CC=C2)C(=O)N1CC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results