missing translation for 'onlineSavingsMsg'
Learn More

cis-1,2-Dicyano-1,2-bis(2,4,5-trimethyl-3-thienyl)ethene 98.0+%, TCI America™

Catalog No. B15361G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15361G 1 g
B15365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15361G Supplier TCI America Supplier No. B15361G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112440-46-7
Molecular Formula C18H18N2S2
Molecular Weight (g/mol) 326.476
MDL Number MFCD00143977
InChI Key AYNDKKQQUZPETC-NXVVXOECSA-N
Synonym 1,2-Bis(2,4,5-trimethyl-3-thienyl)-cis-1,2-dicyanoethene
PubChem CID 44630141
IUPAC Name (Z)-2,3-bis(2,4,5-trimethylthiophen-3-yl)but-2-enedinitrile
SMILES CC1=C(SC(=C1C(=C(C#N)C2=C(SC(=C2C)C)C)C#N)C)C

Specifications

Melting Point 121°C
Quantity 1 g
UN Number 3439
Formula Weight 326.48
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material cis-1,2-Dicyano-1,2-bis(2,4,5-trimethyl-3-thienyl)ethene