accessibility menu, dialog, popup

UserName

CH3SNSCH3H3CCH3NH3CCH3(E/Z)

CAS RN 112440-46-7

IUPAC Name: bis(2,4,5-trimethylthiophen-3-yl)but-2-enedinitrile
Synonyms: bis(2,4,5-trimethylthiophen-3-yl)but-2-enedinitrile
Molecular Weight (g/mol): 326.48
Molecular Formula: C18H18N2S2
InChi Key: AYNDKKQQUZPETC-UHFFFAOYSA-N
SMILES: CC1=C(C)C(C(C#N)=C(C#N)C2=C(C)SC(C)=C2C)=C(C)S1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results