missing translation for 'onlineSavingsMsg'
Learn More

cis-1,2-Cyclohexanedimethanol 97.0+%, TCI America™

Catalog No. C23185G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23185G 5 g
C23181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23185G Supplier TCI America Supplier No. C23185G

Chemical Identifiers

CAS 15753-50-1
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00062987,MFCD16660860,MFCD22572336,MFCD19300921,MFCD22572336,MFCD09260493
InChI Key XDODWINGEHBYRT-UHFFFAOYNA-N
Synonym cis-1,2-Bis(hydroxymethyl)cyclohexane
PubChem CID 2724019
IUPAC Name [2-(hydroxymethyl)cyclohexyl]methanol
SMILES OCC1CCCCC1CO

Specifications

Melting Point 40°C
Color White-Yellow
Boiling Point 112°C
Quantity 5 g
Formula Weight 144.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material cis-1,2-Cyclohexanedimethanol
EINECSNumber (3)-2326
TSCA No