CAS RN 15753-50-1
IUPAC Name:
[2-(hydroxymethyl)cyclohexyl]methanol
Synonyms:
[2-(hydroxymethyl)cyclohexyl]methanol
Molecular Weight (g/mol):
144.21
Molecular Formula:
C8H16O2
InChi Key:
XDODWINGEHBYRT-UHFFFAOYNA-N
SMILES:
OCC1CCCCC1CO
1
–
1
of
1
results
1
cis-1,2-Cyclohexanedimethanol 97.0+%, TCI America™
CAS: 15753-50-1 Molecular Formula: C8H16O2 Molecular Weight (g/mol): 144.21 MDL Number: MFCD00062987,MFCD16660860,MFCD22572336,MFCD19300921,MFCD22572336,MFCD09260493 InChI Key: XDODWINGEHBYRT-UHFFFAOYNA-N Synonym: cis-1,2-Bis(hydroxymethyl)cyclohexane PubChem CID: 2724019 IUPAC Name: [2-(hydroxymethyl)cyclohexyl]methanol SMILES: OCC1CCCCC1CO