missing translation for 'onlineSavingsMsg'
Learn More

cis-1,2-Cyclohexanedicarboxylic anhydride, 99%, Thermo Scientific™

Chemical Identifiers

CAS 13149-00-3
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.17
MDL Number MFCD00005926
InChI Key MUTGBJKUEZFXGO-OLQVQODUSA-N
Synonym cis-hexahydroisobenzofuran-1,3-dione, cis-1,2-cyclohexanedicarboxylic anhydride, cis-hexahydrophthalic anhydride, unii-b52z7tnv87, cis-cyclohexane-1,2-dicarboxylic anhydride, hexahydrophthalic anhydride, cis, 3ar,7as-hexahydro-2-benzofuran-1,3-dione, 1,3-isobenzofurandione, hexahydro-, 3ar,7as-rel, 3ar,7as-hexahydroisobenzofuran-1,3-dione, 1,3-isobenzofurandione, hexahydro-, cis
PubChem CID 641660
IUPAC Name (3aS,7aR)-3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione
SMILES C1CCC2C(C1)C(=O)OC2=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC154032500
SDS View Documents
Thermo Scientific Chemicals
154032500
250g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC154030010
SDS View Documents
Thermo Scientific Chemicals
154030010
1kg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC154030025
SDS View Documents
Thermo Scientific Chemicals
154030025
2.5kg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

13149-00-3
154.17
MUTGBJKUEZFXGO-OLQVQODUSA-N
641660
C1CCC2C(C1)C(=O)OC2=O
C8H10O3
MFCD00005926
cis-hexahydroisobenzofuran-1,3-dione, cis-1,2-cyclohexanedicarboxylic anhydride, cis-hexahydrophthalic anhydride, unii-b52z7tnv87, cis-cyclohexane-1,2-dicarboxylic anhydride, hexahydrophthalic anhydride, cis, 3ar,7as-hexahydro-2-benzofuran-1,3-dione, 1,3-isobenzofurandione, hexahydro-, 3ar,7as-rel, 3ar,7as-hexahydroisobenzofuran-1,3-dione, 1,3-isobenzofurandione, hexahydro-, cis
(3aS,7aR)-3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione

Specifications

13149-00-3
143°C
98.5% min. (GC)
C8H10O3
250g
Solubility in water: hydrolyzes. Other solubilities: miscible with benzene, toluene, acetone, ccl4,, chloroform, ethanol, ethyl acetate, slightly soluble in petroleum ether
C1CCC2C(C1)C(=O)OC2=O
154.17
26 mPa.s (50°C)
99%
296.0°C
Authentic
Glass bottle
MFCD00005926
cis-hexahydroisobenzofuran-1,3-dione, cis-1,2-cyclohexanedicarboxylic anhydride, cis-hexahydrophthalic anhydride, unii-b52z7tnv87, cis-cyclohexane-1,2-dicarboxylic anhydride, hexahydrophthalic anhydride, cis, 3ar,7as-hexahydro-2-benzofuran-1,3-dione, 1,3-isobenzofurandione, hexahydro-, 3ar,7as-rel, 3ar,7as-hexahydroisobenzofuran-1,3-dione, 1,3-isobenzofurandione, hexahydro-, cis
MUTGBJKUEZFXGO-OLQVQODUSA-N
(3aS,7aR)-3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione
641660
154.17
cis-1, 2-Cyclohexanedicarboxylic anhydride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 236-086-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only