Learn More
cis-1,2-Cyclohexanedicarboxylic anhydride, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 154030010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| cis-1, 2-Cyclohexanedicarboxylic anhydride | |
| 296.0°C | |
| Authentic | |
| Glass bottle | |
| MFCD00005926 | |
| cis-hexahydroisobenzofuran-1,3-dione, cis-1,2-cyclohexanedicarboxylic anhydride, cis-hexahydrophthalic anhydride, unii-b52z7tnv87, cis-cyclohexane-1,2-dicarboxylic anhydride, hexahydrophthalic anhydride, cis, 3ar,7as-hexahydro-2-benzofuran-1,3-dione, 1,3-isobenzofurandione, hexahydro-, 3ar,7as-rel, 3ar,7as-hexahydroisobenzofuran-1,3-dione, 1,3-isobenzofurandione, hexahydro-, cis | |
| MUTGBJKUEZFXGO-OLQVQODUSA-N | |
| (3aS,7aR)-3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione | |
| 641660 | |
| 154.17 |
| 13149-00-3 | |
| 143°C | |
| 98.5% min. (GC) | |
| C8H10O3 | |
| 1kg | |
| Solubility in water: hydrolyzes. Other solubilities: miscible with benzene, toluene, acetone, ccl4,, chloroform, ethanol, ethyl acetate, slightly soluble in petroleum ether | |
| C1CCC2C(C1)C(=O)OC2=O | |
| 154.17 | |
| 26 mPa.s (50°C) | |
| 99% |
Chemical Identifiers
| 13149-00-3 | |
| 154.17 | |
| MUTGBJKUEZFXGO-OLQVQODUSA-N | |
| 641660 | |
| C1CCC2C(C1)C(=O)OC2=O |
| C8H10O3 | |
| MFCD00005926 | |
| cis-hexahydroisobenzofuran-1,3-dione, cis-1,2-cyclohexanedicarboxylic anhydride, cis-hexahydrophthalic anhydride, unii-b52z7tnv87, cis-cyclohexane-1,2-dicarboxylic anhydride, hexahydrophthalic anhydride, cis, 3ar,7as-hexahydro-2-benzofuran-1,3-dione, 1,3-isobenzofurandione, hexahydro-, 3ar,7as-rel, 3ar,7as-hexahydroisobenzofuran-1,3-dione, 1,3-isobenzofurandione, hexahydro-, cis | |
| (3aS,7aR)-3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 236-086-3
RUO â Research Use Only