missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-1,2-Cyclohexanediamine 97.0+%, TCI America™
Supplier: TCI America C146325G
Specifications
| cis-1,2-Cyclohexanediamine | |
| Yellow | |
| C6H14N2 | |
| 25 g | |
| cis-1,2-diaminocyclohexane, cis-1,2-cyclohexanediamine, 1r,2s-cyclohexane-1,2-diamine, unii-r0zn6k26ep, cis-cyclohexane-1,2-diamine, 1,2-diaminocyclohexane, cis, r0zn6k26ep, 1,2-cyclohexanediamine, 1r,2s-rel, 1,2-cyclohexanediamine #, cis-1,2-cyclohexandiamine | |
| NC1CCCCC1N | |
| 114.19 | |
| 114.19 | |
| Liquid |
| 1436-59-5 | |
| 93°C | |
| MFCD00063746 | |
| 2735 | |
| SSJXIUAHEKJCMH-UHFFFAOYNA-N | |
| cyclohexane-1,2-diamine | |
| 342917 | |
| ≥97.0% (GC,T) |
Chemical Identifiers
| 1436-59-5 | |
| 114.19 | |
| SSJXIUAHEKJCMH-UHFFFAOYNA-N | |
| 342917 | |
| NC1CCCCC1N |
| C6H14N2 | |
| MFCD00063746 | |
| cis-1,2-diaminocyclohexane, cis-1,2-cyclohexanediamine, 1r,2s-cyclohexane-1,2-diamine, unii-r0zn6k26ep, cis-cyclohexane-1,2-diamine, 1,2-diaminocyclohexane, cis, r0zn6k26ep, 1,2-cyclohexanediamine, 1r,2s-rel, 1,2-cyclohexanediamine #, cis-1,2-cyclohexandiamine | |
| cyclohexane-1,2-diamine |
Safety and Handling
EINECSNumber : (3)-4230
TSCA : No