missing translation for 'onlineSavingsMsg'
Learn More

cis-1,2-Cyclohexanediamine 97.0+%, TCI America™

Catalog No. p-7124998
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14635G 5 g
C146325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14635G Supplier TCI America Supplier No. C14635G
Only null left

Chemical Identifiers

CAS 1436-59-5
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.19
MDL Number MFCD00063746
InChI Key SSJXIUAHEKJCMH-UHFFFAOYNA-N
Synonym cis-1,2-diaminocyclohexane, cis-1,2-cyclohexanediamine, 1r,2s-cyclohexane-1,2-diamine, unii-r0zn6k26ep, cis-cyclohexane-1,2-diamine, 1,2-diaminocyclohexane, cis, r0zn6k26ep, 1,2-cyclohexanediamine, 1r,2s-rel, 1,2-cyclohexanediamine #, cis-1,2-cyclohexandiamine
PubChem CID 342917
IUPAC Name cyclohexane-1,2-diamine
SMILES NC1CCCCC1N

Specifications

Color Yellow
Boiling Point 93°C
Quantity 5 g
UN Number 2735
Formula Weight 114.19
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material cis-1,2-Cyclohexanediamine
EINECSNumber (3)-4230
TSCA No