missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-1,2,3,6-Tetrahydrophthalimide 98.0+%, TCI America™
Supplier: TCI America T2801250G
Specifications
| cis-1,2,3,6-Tetrahydrophthalimide | |
| 138°C | |
| MFCD00005880 | |
| cis-1,2,3,6-tetrahydrophthalimide, cis-3a,4,7,7a-tetrahydro-1h-isoindole-1,3 2h-dione, cis-4-cyclohexene-1,2-dicarboximide, cis-tetrahydrophthalimide, unii-es57d28o1u, 4-cyclohexene-1,2-dicarboximide, cis, cis-.delta.4-tetrahydrophthalimide, 1h-isoindole-1,3 2h-dione, 3a,4,7,7a-tetrahydro-, cis, 1h-isoindole-1,3 2h-dione, 3a,4,7,7a-tetrahydro-, 3ar,7as-rel, 3ar,7as-3a,4,7,7a-tetrahydroisoindole-1,3-dione | |
| C1C=CCC2C1C(=O)NC2=O | |
| 151.165 | |
| 151.17 | |
| Crystalline Powder |
| 1469-48-3 | |
| C8H9NO2 | |
| 250 g | |
| CIFFBTOJCKSRJY-OLQVQODUSA-N | |
| (3aR,7aS)-3a,4,7,7a-tetrahydroisoindole-1,3-dione | |
| 92888 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 1469-48-3 | |
| 151.165 | |
| CIFFBTOJCKSRJY-OLQVQODUSA-N | |
| 92888 | |
| C1C=CCC2C1C(=O)NC2=O |
| C8H9NO2 | |
| MFCD00005880 | |
| cis-1,2,3,6-tetrahydrophthalimide, cis-3a,4,7,7a-tetrahydro-1h-isoindole-1,3 2h-dione, cis-4-cyclohexene-1,2-dicarboximide, cis-tetrahydrophthalimide, unii-es57d28o1u, 4-cyclohexene-1,2-dicarboximide, cis, cis-.delta.4-tetrahydrophthalimide, 1h-isoindole-1,3 2h-dione, 3a,4,7,7a-tetrahydro-, cis, 1h-isoindole-1,3 2h-dione, 3a,4,7,7a-tetrahydro-, 3ar,7as-rel, 3ar,7as-3a,4,7,7a-tetrahydroisoindole-1,3-dione | |
| (3aR,7aS)-3a,4,7,7a-tetrahydroisoindole-1,3-dione |
Safety and Handling
EINECSNumber : (5)-3359
TSCA : No