missing translation for 'onlineSavingsMsg'
Learn More

cis-1,2,3,6-Tetrahydrophthalimide 98.0+%, TCI America™

Catalog No. p-7137426
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T280125G 25 g
T2801250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T280125G Supplier TCI America Supplier No. T280125G

Chemical Identifiers

CAS 1469-48-3
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00005880
InChI Key CIFFBTOJCKSRJY-OLQVQODUSA-N
Synonym cis-1,2,3,6-tetrahydrophthalimide, cis-3a,4,7,7a-tetrahydro-1h-isoindole-1,3 2h-dione, cis-4-cyclohexene-1,2-dicarboximide, cis-tetrahydrophthalimide, unii-es57d28o1u, 4-cyclohexene-1,2-dicarboximide, cis, cis-.delta.4-tetrahydrophthalimide, 1h-isoindole-1,3 2h-dione, 3a,4,7,7a-tetrahydro-, cis, 1h-isoindole-1,3 2h-dione, 3a,4,7,7a-tetrahydro-, 3ar,7as-rel, 3ar,7as-3a,4,7,7a-tetrahydroisoindole-1,3-dione
PubChem CID 92888
IUPAC Name (3aR,7aS)-3a,4,7,7a-tetrahydroisoindole-1,3-dione
SMILES C1C=CCC2C1C(=O)NC2=O

Specifications

Melting Point 138°C
Quantity 25 g
Formula Weight 151.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material cis-1,2,3,6-Tetrahydrophthalimide
EINECSNumber (5)-3359
TSCA No