missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Circulation Tubing Kit

Catalog No. 16200484
Change view
Click to view available options
Diameter (English) Inside:
0.75 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English) Inside
16200484 0.75 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16200484 Supplier Labconco™ Supplier No. 5441808

Connects AtmoSense Oxygen and Moisture Monitors in-line between the Protector Controlled Atmosphere or Combination Glove Box or the Precise Controlled Atmosphere Glove Box and the AtmosPure Re-Gen Gas Purifier.

  • Two 2cm (0.75 in.) I. D. flexible neoprene tubes
  • Brass fittings 2cm (0.75 in.) O.D. on each end of tubing
  • Includes two sections of tubing with fitting

Specifications

Product Type Tubing Kit
Diameter (English) Inside 0.75 in.