missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CIRBP Recombinant Rabbit Monoclonal Antibody (23GB7980)

Catalog No. MA553307
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553307 20 μL
MA553306 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553307 Supplier Invitrogen™ Supplier No. MA553307
Only null left

Rabbit Recombinant Monoclonal Antibody

CIRBP Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

CIRBP is cold-inducible mRNA binding protein that plays a protective role in the genotoxic stress response by stabilizing transcripts of genes involved in cell survival. It acts as a translational activator and seems to play an essential role in cold-induced suppression of cell proliferation. It binds specifically to the 3'-untranslated regions (3'-UTRs) of stress-responsive transcripts RPA2 and TXN. This protein acts as a translational repressor and promotes assembly of stress granules (SGs), when overexpressed.

Specifications

Antigen CIRBP
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB7980
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CIRBP
Gene Accession No. Q14011
Gene Alias A18 hnRNP; A18HNRNP; CIRBP; CIRP; cold inducible RNA binding protein; cold inducible RNA-binding protein; cold-inducible RNA-binding protein; glycine-rich RNA binding protein; glycine-rich RNA-binding protein CIRP; R74941; testicular tissue protein Li 39
Gene Symbols CIRBP
Host Species Rabbit
Immunogen A synthesized peptide derived from CIRBP (1-172AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1153
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less