missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CIR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568972
Description
This target displays homology in the following species: Human: 100%.
CIR1 is a protein coding gene.Chemical Identifiers
| CIR1 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| CIR1 | |
| 1700023b02rik; 2810021A19Rik; CBF1 (RBPJ) interacting corepressor 1; CBF1 interacting corepressor; CBF1-interacting corepressor; Cicr; Cir; CIR1; Corepressor interacting with RBPJ 1; corepressor interacting with RBPJ, 1; corepressor interacting with RBPJ, CIR1; recepin; RGD1309199 | |
| CIR1 | |
| Primary | |
| Affinity chromatography |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q86X95 | |
| 9541 | |
| synthetic peptide directed towards the C terminal region of human CIR. | |
| Antibody |
Specifications
| CIR1 | |
| Polyclonal | |
| Unconjugated | |
| CIR1 | |
| 1700023b02rik; 2810021A19Rik; CBF1 (RBPJ) interacting corepressor 1; CBF1 interacting corepressor; CBF1-interacting corepressor; Cicr; Cir; CIR1; Corepressor interacting with RBPJ 1; corepressor interacting with RBPJ, 1; corepressor interacting with RBPJ, CIR1; recepin; RGD1309199 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 9541 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q86X95 | |
| CIR1 | |
| synthetic peptide directed towards the C terminal region of human CIR. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |