missing translation for 'onlineSavingsMsg'
Learn More

Ciprofloxacin Hydrochloride, MP Biomedicals™

Catalog No. MP219902050
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219902050 50 g
MP219902005 5 g
MP219902025 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP219902050 Supplier MP Biomedicals Inc Supplier No. 0219902050
Only null left

Very effective against a broad spectrum of bacteria and mycoplasma. Ciprofloxacin Hydrochloride, MP Biomedicals is a fluorinated quinolone antibacterial agent. Typically used for 12–20 days in cell culture media, with the media being changed every 3

During the proliferation phase of a bacterium, a segmental twisting and untwisting of the chromosomes takes place. An enzyme called DNA gyrase plays a decisive part in this process. Ciprofloxacin inhibits this DNA gyrase in a way that arrests the bacterial metabolism, since vital information can no longer be read from the bacterial chromosome. Ultimately, it affects DNA replication and cell division.

Ciprofloxacin is most commonly used to treat infections of the skin, bone, sinus, lung, abdomen, and bladder. It can also be used to treat some sexually transmitted infections. Additionally, ciprofloxacin can be used after exposure to inhalation of anthrax to reduce the chance of developing an infection.

Chemical Identifiers

CAS 86393-32-0
Molecular Formula C17H21ClFN3O4
Molecular Weight (g/mol) 385.82
MDL Number MFCD00242856
InChI Key ARPUHYJMCVWYCZ-UHFFFAOYSA-N
Synonym Cipro, Ciproxan, 1-Cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperzinyl)-3-quinolinecarboxylic acid HCl H2O
PubChem CID 62998
ChEBI CHEBI:59936
IUPAC Name hydrogen 1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrate chloride
SMILES [H+].O.[Cl-].OC(=O)C1=CN(C2CC2)C2=CC(N3CCNCC3)=C(F)C=C2C1=O

Specifications

Physical Form Powder
pH 3.7 (2% aq. soln.) (literature)
Melting Point 313°C to 322°C
Quantity 50 g
Solubility Information Stock solutions can be prepared in water or 0.1M NaOH (1-10mg/mL).
Formula Weight 367.8g/mol (anhyd.)
Recommended Storage Store at 4°C.