missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ciprofloxacin hydrochloride monohydrate
Description
Ciprofloxacin hydrochloride has been suggested as a treatment for a wide range of infections caused by susceptible organisms including infections of urinary, respiratory, and gastrointestinal tracts, gonorrhoea and septicaemia. Nalidixic acid analogue.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ciprofloxacin hydrochloride has been suggested as a treatment for a wide range of infections caused by susceptible organisms including infections of urinary, respiratory, and gastrointestinal tracts, gonorrhoea and septicaemia. May be given by mouth or may be administered by intravenous infusion. Nalidixic acid analogue.
Solubility
Soluble in water (35mg/ml).
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Ciprofloxacin hydrochloride has been suggested as a treatment for a wide range of infections caused by susceptible organisms including infections of urinary, respiratory, and gastrointestinal tracts, gonorrhoea and septicaemia. May be given by mouth or may be administered by intravenous infusion. Nalidixic acid analogue.
Solubility
Soluble in water (35mg/ml).
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Specifications
Specifications
| Type | Ciprofloxacin Hydrochloride |
| Physical Form | Lyophilized Powder |
| Color | White/Yellow |
| Quantity | 25 g |
| Merck Index | 14,2314 |
| Solubility Information | Soluble in water (35mg/ml). |
| Formula Weight | 385.82 (367.80 Anhydrous) |
| Chemical Name or Material | Ciprofloxacin hydrochloride |
Safety and Handling
| RTECSNumber | VB1993800 |
| TSCA | No |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only