missing translation for 'onlineSavingsMsg'
Learn More

Ciprofloxacin Hydrochloride Monohydrate 98.0+%, TCI America™

Supplier:  TCI America C222725G

 View more versions of this product

Catalog No. C222725G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ciprofloxacin Hydrochloride Monohydrate
White-Yellow
MFCD00242856
ciprofloxacin hydrochloride monohydrate, ciloxan, ceprimax, oftacilox, proquin xr, cipro, floxacipron, belmacina, ciflosin, ciprocinal
[H+].O.[Cl-].OC(=O)C1=CN(C2CC2)C2=CC(N3CCNCC3)=C(F)C=C2C1=O
385.82
CHEBI:59936
≥98.0% (HPLC,T)
86393-32-0
C17H21ClFN3O4
25 g
ARPUHYJMCVWYCZ-UHFFFAOYSA-N
hydrogen 1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrate chloride
62998
367.81
Crystalline Powder

Chemical Identifiers

86393-32-0
385.82
ARPUHYJMCVWYCZ-UHFFFAOYSA-N
62998
hydrogen 1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrate chloride
C17H21ClFN3O4
MFCD00242856
ciprofloxacin hydrochloride monohydrate, ciloxan, ceprimax, oftacilox, proquin xr, cipro, floxacipron, belmacina, ciflosin, ciprocinal
CHEBI:59936
[H+].O.[Cl-].OC(=O)C1=CN(C2CC2)C2=CC(N3CCNCC3)=C(F)C=C2C1=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions