missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ciprofloxacin Hydrochloride Monohydrate 98.0+%, TCI America™
Supplier: TCI America C2227100G
Specifications
| Ciprofloxacin Hydrochloride Monohydrate | |
| White-Yellow | |
| MFCD00242856 | |
| ciprofloxacin hydrochloride monohydrate, ciloxan, ceprimax, oftacilox, proquin xr, cipro, floxacipron, belmacina, ciflosin, ciprocinal | |
| [H+].O.[Cl-].OC(=O)C1=CN(C2CC2)C2=CC(N3CCNCC3)=C(F)C=C2C1=O | |
| 385.82 | |
| CHEBI:59936 | |
| ≥98.0% (HPLC,T) |
| 86393-32-0 | |
| C17H21ClFN3O4 | |
| 100 g | |
| ARPUHYJMCVWYCZ-UHFFFAOYSA-N | |
| hydrogen 1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrate chloride | |
| 62998 | |
| 367.81 | |
| Crystalline Powder |
Chemical Identifiers
| 86393-32-0 | |
| 385.82 | |
| ARPUHYJMCVWYCZ-UHFFFAOYSA-N | |
| 62998 | |
| hydrogen 1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrate chloride |
| C17H21ClFN3O4 | |
| MFCD00242856 | |
| ciprofloxacin hydrochloride monohydrate, ciloxan, ceprimax, oftacilox, proquin xr, cipro, floxacipron, belmacina, ciflosin, ciprocinal | |
| CHEBI:59936 | |
| [H+].O.[Cl-].OC(=O)C1=CN(C2CC2)C2=CC(N3CCNCC3)=C(F)C=C2C1=O |
Safety and Handling
TSCA : No