missing translation for 'onlineSavingsMsg'
Learn More

Ciprofloxacin Hydrochloride Monohydrate 98.0+%, TCI America™

Catalog No. C22275G
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22275G 5 g
C222725G 25 g
C2227100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C22275G Supplier TCI America Supplier No. C22275G
Only null left

Chemical Identifiers

CAS 86393-32-0
Molecular Formula C17H21ClFN3O4
Molecular Weight (g/mol) 385.82
MDL Number MFCD00242856
InChI Key ARPUHYJMCVWYCZ-UHFFFAOYSA-N
Synonym ciprofloxacin hydrochloride monohydrate, ciloxan, ceprimax, oftacilox, proquin xr, cipro, floxacipron, belmacina, ciflosin, ciprocinal
PubChem CID 62998
ChEBI CHEBI:59936
IUPAC Name hydrogen 1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrate chloride
SMILES [H+].O.[Cl-].OC(=O)C1=CN(C2CC2)C2=CC(N3CCNCC3)=C(F)C=C2C1=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 367.81
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ciprofloxacin Hydrochloride Monohydrate