missing translation for 'onlineSavingsMsg'
Learn More

STERIS™ CIP 220 Acid-Based Process and Research Cleaner

CIP 220 Acid-Based Cleaner: A versatile, low-foaming, phosphate-free detergent containing hydroxyacetic (glycolic) acid. Ideal for pharmaceutical cleaning in CIP, COP, and manual applications. Validates easily and supports global harmonization. pH - (1%) 2.8, typical.

$857.06 - $42,318.26

Specifications

Product Type Acid-Based Cleaner
Color-coded No
Form Clear Liquid
pH 2.8 (1%)
Scent Unscented
View More Specs

Products 3
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17355263
View Documents Promotion Details
STERIS™
1D2208
4 x 1 gal.
Case of 4 Each for $857.06
Only null left
 
17355266
View Documents Promotion Details
STERIS™
1D2201
55 gal.
Each for $8,536.63
Only null left
 
17355262
View Documents Promotion Details
STERIS™
1D22D3
300 gal. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

CIP 220 Acid-Based Process and Research Cleaner is specially formulated to meet the unique cleaning demands found in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable cleaner.

HOW CIP 220 WORKS

CIP 220 Acid Detergent is a proprietary blend of hydroxyacetic (glycolic) acid, an advanced surfactant system and other performance-enhancing ingredients that provide multiple cleaning mechanisms. This phosphate free low foaming detergent is ideal for use in CIP, COP and manual applications.

WHY CIP 220?

  • Flexibility - Effective cleaning against a wide variety of pharmaceutical process residues in clean-in-place (CIP), clean-out-of-place (COP), and manual applications.
  • Easy to Validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports.
  • Cost Savings - Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics.
  • Globally Availability - phosphate free, and manufactured with biodegradable surfactants to support harmonization of cleaning processes.
  • Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility.
Specifications

Specifications

Acid-Based Cleaner
Clear Liquid
Unscented
No
2.8 (1%)
Non-sterile
Videos
SDS
Documents

Documents