missing translation for 'onlineSavingsMsg'
Learn More

STERIS™ CIP 220 Acid-Based Process and Research Cleaner

Catalog No. 17355266
Change view
Click to view available options
Size:
4 x 1 gal.
55 gal.
300 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17355266 55 gal.
17355263 4 x 1 gal.
17355262 300 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17355266 Supplier STERIS™ Supplier No. 1D2201
Only null left

CIP 220 Acid-Based Cleaner: A versatile, low-foaming, phosphate-free detergent containing hydroxyacetic (glycolic) acid. Ideal for pharmaceutical cleaning in CIP, COP, and manual applications. Validates easily and supports global harmonization. pH - (1%) 2.8, typical.

CIP 220 Acid-Based Process and Research Cleaner is specially formulated to meet the unique cleaning demands found in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable cleaner.

HOW CIP 220 WORKS

CIP 220 Acid Detergent is a proprietary blend of hydroxyacetic (glycolic) acid, an advanced surfactant system and other performance-enhancing ingredients that provide multiple cleaning mechanisms. This phosphate free low foaming detergent is ideal for use in CIP, COP and manual applications.

WHY CIP 220?

  • Flexibility - Effective cleaning against a wide variety of pharmaceutical process residues in clean-in-place (CIP), clean-out-of-place (COP), and manual applications.
  • Easy to Validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports.
  • Cost Savings - Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics.
  • Globally Availability - phosphate free, and manufactured with biodegradable surfactants to support harmonization of cleaning processes.
  • Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility.

Specifications

Product Type Acid-Based Cleaner
Color-coded No
Form Clear Liquid
pH 2.8 (1%)
Scent Unscented
Sterility Non-sterile
Size 55 gal.