missing translation for 'onlineSavingsMsg'
Learn More

Cinnarizine

Catalog No. AAJ6456814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6456814 25 g
AAJ6456822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6456814 Supplier Thermo Scientific Chemicals Supplier No. J6456814

An antihistamine | CAS: 298-57-7 | C26H28N2 | 368.524 g/mol

An antihistamine

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 298-57-7
Molecular Formula C26H28N2
Molecular Weight (g/mol) 368.524
MDL Number MFCD00056037
InChI Key DERZBLKQOCDDDZ-JLHYYAGUSA-N
Synonym cinnarizine, stugeron, dimitronal, stutgin, dimitron, folcodal, midronal, mitronal, stutgeron, glanil
PubChem CID 1547484
ChEBI CHEBI:31403
IUPAC Name 1-benzhydryl-4-[(E)-3-phenylprop-2-enyl]piperazine
SMILES C1CN(CCN1CC=CC2=CC=CC=C2)C(C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 117°C to 121°C
Color White to Yellow
Quantity 25 g
Merck Index 14,2305
Solubility Information Soluble in chloroform at 50mg/ml
Formula Weight 368.52
Physical Form Powder
Chemical Name or Material Cinnarizine
EINECSNumber 206-064-8
RTECSNumber TL3430000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only