missing translation for 'onlineSavingsMsg'
Learn More

Cinnarizine 98.0+%, TCI America™

Catalog No. p-7125965
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
50 g
Catalog No. Quantity
C345910G 10 g
C345950G 50 g
2 options

Catalog No. C345910G

Supplier: TCI America C345910G

Only null left

Chemical Identifiers

CAS 298-57-7
Molecular Formula C26H28N2
Molecular Weight (g/mol) 368.524
MDL Number MFCD00056037
InChI Key DERZBLKQOCDDDZ-JLHYYAGUSA-N
Synonym cinnarizine, stugeron, dimitronal, stutgin, dimitron, folcodal, midronal, mitronal, stutgeron, glanil
PubChem CID 1547484
ChEBI CHEBI:31403
IUPAC Name 1-benzhydryl-4-[(E)-3-phenylprop-2-enyl]piperazine
SMILES C1CN(CCN1CC=CC2=CC=CC=C2)C(C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 119°C
Color White-Yellow
Quantity 10 g
Formula Weight 368.52
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Cinnarizine
EINECSNumber (5)-0960
TSCA No
Recommended Storage Refrigerator