missing translation for 'onlineSavingsMsg'
Learn More

Cinnamylideneacetophenone, 98+%

CAS: 614-57-3 | C17H14O | 234.298 g/mol

$41.32 - $140.18

Chemical Identifiers

CAS 614-57-3
Molecular Formula C17H14O
Molecular Weight (g/mol) 234.298
MDL Number MFCD00020692
InChI Key QONKLJMPKWQQFG-HPIZBCMHSA-N
Synonym cinnamylideneacetophenone, cinnamalacetophenone, 5-phenyl-2,4-pentadienophenone, 5-phenylpenta-2,4-dienophenone, 2e,4e-1,5-diphenylpenta-2,4-dien-1-one, ?,e-1,5-diphenyl-2,4-pentadien-1-one, 2,4-pentadienophenone, 5-phenyl, 2,4-pentadien-1-one, 1,5-diphenyl-, ?,e, 1,5-diphenyl-2,4-pentadien-1-one, 2,4-pentadien-1-one, 1,5-diphenyl
PubChem CID 1549519
IUPAC Name (2E,4E)-1,5-diphenylpenta-2,4-dien-1-one
SMILES C1=CC=C(C=C1)C=CC=CC(=O)C2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1511406
View Documents
Thermo Scientific Chemicals
A1511406
5 g N/A
Only null left
 
AAA1511414
View Documents
Thermo Scientific Chemicals
A1511414
25 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

614-57-3
234.298
QONKLJMPKWQQFG-HPIZBCMHSA-N
1549519
C1=CC=C(C=C1)C=CC=CC(=O)C2=CC=CC=C2
C17H14O
MFCD00020692
cinnamylideneacetophenone, cinnamalacetophenone, 5-phenyl-2,4-pentadienophenone, 5-phenylpenta-2,4-dienophenone, 2e,4e-1,5-diphenylpenta-2,4-dien-1-one, ?,e-1,5-diphenyl-2,4-pentadien-1-one, 2,4-pentadienophenone, 5-phenyl, 2,4-pentadien-1-one, 1,5-diphenyl-, ?,e, 1,5-diphenyl-2,4-pentadien-1-one, 2,4-pentadien-1-one, 1,5-diphenyl
(2E,4E)-1,5-diphenylpenta-2,4-dien-1-one

Specifications

614-57-3
C17H14O
5 g
cinnamylideneacetophenone, cinnamalacetophenone, 5-phenyl-2,4-pentadienophenone, 5-phenylpenta-2,4-dienophenone, 2e,4e-1,5-diphenylpenta-2,4-dien-1-one, ?,e-1,5-diphenyl-2,4-pentadien-1-one, 2,4-pentadienophenone, 5-phenyl, 2,4-pentadien-1-one, 1,5-diphenyl-, ?,e, 1,5-diphenyl-2,4-pentadien-1-one, 2,4-pentadien-1-one, 1,5-diphenyl
C1=CC=C(C=C1)C=CC=CC(=O)C2=CC=CC=C2
234.298
234.3
Cinnamylideneacetophenone
100°C to 102°C
MFCD00020692
1870052
QONKLJMPKWQQFG-HPIZBCMHSA-N
(2E,4E)-1,5-diphenylpenta-2,4-dien-1-one
1549519
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 210-385-9

RTECSNumber : RZ2477000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only